missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GP25L2 Protein

A DNA sequence encoding the Homo sapiens (Human) GP25L2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8481-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064851

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_059980.2
GP25L2 Protein
Research Use Only
GP25L2
Recombinant Protein
E. coli
FHIGETEKKC FIEEIPDETM VIGNYRTQLY DKQREEYQPA TPGLGMFVEV KDPEDKVILA RQYGSEGRFT FTSHTPGEHQ ICLHSNSTKF SLFAGGMLRV HLDIQVGEHA NDYAEIAAKD KLSELQLRVR QLVEQVEQIQ KEQNYQRWRE ERFRQTSE
Greater than 90% as determined by SDS-PAGE.
54732
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers