missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human gp130 His-tag Avi-tag Protein
Description
Measured by its binding ability in a functional ELISA. When Biotinylated Recombinant Human gp130 His-tag Avi-tag is captured on a Streptavidin Coated Plate (Catalog # CP004 ), it binds to Recombinant Human IL-6 (Catalog # 7270-IL ) in the presence of Recombinant Human IL-6R alpha (Catalog # 227-SR ). The ED50 for this binding is 0.800-4.80 ng/mL.
Specifications
Specifications
| Accession Number | AAA59155.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3572 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 93-103 kDa, under reducing conditions.; MolecularWeight-theroretical: 70 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human gp130 protein Human gp130 (Glu23-Ile618) (N-terminus) Accession # AAA59155.1 6-His tag Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6 R subunit beta, IL-6 RB, IL-6 R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?