missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAZ Protein
A cDNA sequence encoding the GNAZ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12012-20ug
Additional Details : Weight : 0.01000kg
Specifications
GNAZ Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGCRQSSEEK EAARRSRRID RHLRSESQRQ RREIKLLLLG TSNSGKSTIV KQMKIIHSGG FNLEACKEYK PLIIYNAIDS LTRIIRALAA LRIDFHNPDR AYDAVQLFAL TGPAESKGEI TPELLGVMRR LWADPGAQAC FSRSSEYHLE DNAAYYLNDL ERIAAADYIP TVEDILRSRD MTTGIVENKF TFKELTFKMV DVGGQRSERK KWIHCFEGVT AIIFCVELSG YDLKLYEDNQ TSRMAESLRL FDSICNNNWF INTSLILFLN KKDLLAEKIR RIPLTICFPE YKGQNTYEEA AVYIQRQFED LNRNKETKEI YSHFTCATDT SNIQFVFDAV TDVIIQNNLK YIGLC | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GNAZ solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |