missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAQ Protein
A cDNA sequence encoding the GNAQ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12011-20ug
Additional Details : Weight : 0.01000kg
Specifications
GNAQ Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTLESIM ACCLSEEAKE ARRINDEIER QLRRDKRDAR RELKLLLLGT GESGKSTFIK QMRIIHGSGY SDEDKRGFTK LVYQNIFTAM QAMIRAMDTL KIPYKYEHNK AHAQLVREVD VEKVSAFENP YVDAIKSLWN DPGIQECYDR RREYQLSDST KYYLNDLDRV ADPAYLPTQQ DVLRVRVPTT GIIEYPFDLQ SVIFRMVDVG GQRSERRKWI HCFENVTSIM FLVALSEYDQ VLVESDNENR MEESKALFRT IITYPWFQNS SVILFLNKKD LLEEKIMYSH LVDYFPEYDG PQRDAQAARE FILKMFVDLN PDSDKIIYSH FTCATDTENI RFVFAAVKDT ILQLNLKEYN LV | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNAQ protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |