missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMPR / GMPR1 Protein

Product Code. 16067670
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16067670

Brand: enQuireBio™ QP7200ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) GMPR / GMPR1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_006868.3
Gene ID (Entrez) 2766
Name GMPR / GMPR1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GMPR
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MPRIDADLKL DFKDVLLRPK RSSLKSRAEV DLERTFTFRN SKQTYSGIPI IVANMDTVGT FEMAAVMSQH SMFTAIHKHY SLDDWKLFAT NHPECLQNVA VSSGSGQNDL EKMTSILEAV PQVKFICLDV ANGYSEHFVE FVKLVRAKFP EHTIMAGNVV TGEMVEELIL SGADIIKVGV GPGSVCTTRT KTGVGYPQLS AVIECADSAH GLKGHIISDG GCTCPGDVAK AFGAGADFVM LGGMFSGHTE CAGEVFERNG RKLKLFYGMS SDTAMNKHAG GVAEYRASEG KTVEVPYKGD VENTILDILG GLRSTCTYVG AAKLKELSRR ATFIRVTQQH NTVFS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado