missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMFB His Protein

A cDNA sequence encoding the GMFB His was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12005-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949068

  • £155.00 / 25µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GMFB His Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSESLVVCDV AEDLVEKLRK FRFRKETNNA AIIMKIDKDK RLVVLDEELE GISPDELKDE LPERQPRFIV YSYKYQHDDG RVSYPLCFIF SSPVGCKPEQ QMMYAGSKNK LVQTAELTKV FEIRNTEDLT EEWLREKLGF FH
Greater than 95.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GMFB 1 mg/ml protein solution contains 20mM Tris-HCL pH 8.0, 1mM DTT, 0.1M NaCl and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers