missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein
A DNA sequence encoding the Homo sapiens (Human) GM-CSF / CSF2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5876-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000749.2 | |
GM-CSF / CSF2 Protein | |
Research Use Only | |
CSF2 | |
Recombinant Protein | |
E. coli | |
APARSPSPST QPWEHVNAIQ EARRLLNLSR DTAAEMNETV EVISEMFDLQ EPTCLQTRLE LYKQGLRGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFESFKENL KDFLLVIPFD CWEPVQE | |
Greater than 90% as determined by SDS-PAGE. |
1437 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |