missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Glypican 3 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
Description
The Recombinant Human Glypican 3 Protein is derived from NS0. The Recombinant Human Glypican 3 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 2719 |
| Molecular Weight (g/mol) | 61.6 kDa |
| Name | Glypican 3 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Glypican 3 protein Met1-His559 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Glypican 3 is subject to endoproteolytic processing by proprotein convertases (PC). By amino acid sequencing, three peptides (the first with a blocked N-terminus most likely starts with Gln25, the second peptide starts with Ser359 after a furin cleavage s |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction