missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Glyoxalase I Protein
Click to view available options
:
2500ug; Carrier Free
Description
The Recombinant Human Glyoxalase I Protein is derived from E. coli. The Recombinant Human Glyoxalase I Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-HCl and DTT. |
| Gene ID (Entrez) | 2739 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | Glyoxalase I |
| Quantity | 2500μg |
| Source | E. coli-derived human Glyoxalase I protein Ala2-Met184,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction