missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glycophorin A / CD235a Protein

A DNA sequence encoding the Homo sapiens (Human) Glycophorin A / CD235a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6135-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051060

  • £384.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001295119.1
Glycophorin A / CD235a Protein
Research Use Only
GYPA
Recombinant Protein
E. coli
SSTTGVAMHT STSSSVTKSY ISSQTNDTHK RDTYAATPRA HEVSEISVRT VYPPEEETGE RVQLAHHFSE PE
Greater than 90% as determined by SDS-PAGE.
2993
100 μg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers