missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycodelin Protein
A DNA sequence encoding the Homo sapiens (Human) Glycodelin, was expressed in the hosts and tags indicated.
£595.00 - £1655.00
Specifications
Accession Number | NP_001018058.1 |
---|---|
Gene ID (Entrez) | 5047 |
Name | Glycodelin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003480
|
enQuireBio™
QP6462-MA-20UG |
20 μg |
£595.00
20µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093470
|
enQuireBio™
QP6462-MA-100UG |
100 μg |
£1655.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013480
|
enQuireBio™
QP6462-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001018058.1 | |
Glycodelin Protein | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5047 | |
Research Use Only | |
PAEP | |
Recombinant Protein | |
Mammal | |
MDIPQTKQDL ELPKLAGTWH SMAMATNNIS LMATLKAPLR VHITSLLPTP EDNLEIVLHR WENNSCVEKK VLGEKTENPK KFKINYTVAN EATLLDTDYD NFLFLCLQDT TTPIQSMMCQ YLARVLVEDD EIMQGFIRAF RPLPRHLWYL LDLKQMEEPC RF | |
Greater than 90% as determined by SDS-PAGE. |