missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Glutathione Peroxidase 3/GPX3 FLAG Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| Accession Number | P22352 |
| Concentration | 0.5 mg/ml |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 0.2um-filtered solution in PBS, pH 7.2 |
| Gene ID (Entrez) | 2878 |
| Molecular Weight (g/mol) | M.W. Theoretical: 30 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.1 EU/μg purified protein (LAL test; Lonza). |
| Gene Alias | EC 1.11.1, EC 1.11.1.9, Extracellular glutathione peroxidase, glutathione peroxidase 3, glutathione peroxidase 3 (plasma), GPx-3, GPx-P, GPXP, GSHPx-3, GSHPx-P, Plasma glutathione peroxidase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?