missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucosidase 2 subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Glucosidase 2 subunit beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8431-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001329.1 | |
Glucosidase 2 subunit beta Protein | |
Research Use Only | |
PRKCSH | |
Recombinant Protein | |
E. coli | |
VEVKRPRGVS LTNHHFYDES KPFTCLDGSA TIPFDQVNDD YCDCKDGSDE PGTAACPNGS FHCTNTGYKP LYIPSNRVND GVCDCCDGTD EYNSGVICEN TCKEKGRKER ESLQQMAEVT REGFRLKKIL IEDWKKAREE KQKKLIELQA GKKSLEDQVE MLRTVKEEAE KPEREAKEQH QKLWEEQLAA AKAQQEQELA ADAFKELDDD MDGTVSVTEL QTHPELDTDG DGALSEAEAQ ALLSGDTQTD ATSFYDRVWA AIRDKYRSEA LPTDLPAPSA PDLTEPKE | |
Greater than 90% as determined by SDS-PAGE. |
5589 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |