missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucose-6-phosphate isomerase Protein
A DNA sequence encoding the Homo sapiens (Human) Glucose-6-phosphate isomerase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8317-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000166.2 | |
Glucose-6-phosphate isomerase Protein | |
Research Use Only | |
GPI | |
Recombinant Protein | |
E. coli | |
AALTRDPQFQ KLQQWYREHR SELNLRRLFD ANKDRFNHFS LTLNTNHGHI LVDYSKNLVT EDVMRMLVDL AKSRGVEAAR ERMFNGEKIN YTEGRAVLHV ALRNRSNTPI LVDGKDVMPE VNKVLDKMKS FCQRVRSGDW KGYTGKTITD VINIGIGGSD LGPLMVTEAL KPYSSGGPRV WYVSNIDGTH IAKTLAQLNP ESSLFIIASK TFTTQETITN AETAKEWFLQ AAKDPSAVAK HFVALSTNTT KVKEFGIDPQ NMFEFWDWVG GRYSLWSAIG LSIALHVGFD NFEQLLSGAH WMDQHFRTTP LEKNAPVLLA LLGIWYINCF GCETHAMLPY DQYLHRFAAY FQQGDMESNG KYITKSGTRV DHQTGPIVWG EPGTNGQHAF YQLIHQGTKM IPCDFLIPVQ TQHPIRKGLH HKILLANFLA QTEALMRGKS TEEARKELQA AGKSPEDLER LLPHKVFEGN RPTNSIVFTK LTPFMLGALV AMYEHKIFVQ GIIWDINSFD QWGVELGKQL AKKIEPELDG SAQVTSHDAS TNGLINFIKQ QRE | |
Greater than 90% as determined by SDS-PAGE. |
2821 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |