missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Glucose 6 phosphate isomerase His Protein

Product Code. 18342362 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342362 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342362 Supplier Novus Biologicals™ Supplier No. NBP307000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH8.0), 1 mM DTT, 10% glycerol
Gene ID (Entrez) 2821
Molecular Weight (g/mol) M.W. Theoretical: 65.3 kDa
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1.0 EU per 1 microgram of protein (determined by LAL method)
Gene Alias AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36
Gene Symbol GPI
Research Category Cytokine Research
Purity or Quality Grade >95%, by SDS-PAGE
Protein Glucose 6 phosphate isomerase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.