missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Glucokinase/GCK Protein

Product Code. 18347467
Change view
Click to view available options
:
20ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18347467 20ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347467 Supplier R&D Systems™ Supplier No. 7840GK020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer phosphate from ATP to glucose. The specific activity is >265 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35557
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Brij-35 and DTT.
Gene ID (Entrez) 2645
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 63 kDa
Name Glucokinase/GCK
Quantity 20 μg
Source E. coli-derived human Glucokinase/GCK protein Met 6-His tag Human Cystatin A (Met1-Phe98) (N-terminus) Accession # P01040 GS Human Glucokinase/GCK (Val16-Gln465) (N-terminus) Accession # P35557 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol GCK
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein Glucokinase/GCK
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.