missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GITR Ligand/TNFSF18 Protein

Product Code. 18318737
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18318737 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 25µg
18390048 Lyophilized from a 0.2 μm filtered solution in PBS. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18318737 Supplier R&D Systems™ Supplier No. 694GL025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A New and Improved rh GITR-L is Available! CHO expressed; ∼70 fold better activity; HA tag!

Measured by its ability to compete with biotinylated recombinant human GITR Ligand for binding to immobilized recombinant human GITR Fc Chimera.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UNG2
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 8995
Molecular Weight (g/mol) M.W. (Observed): 14 kDa-16 kDa, reducing conditions; M.W. (theoretical): 15 kDa
Name GITR Ligand/TNFSF18
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human GITR Ligand/TNFSF18 protein Glu74-Ser199, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbol TNFSF18
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein GITR Ligand/TNFSF18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.