missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GITR Ligand Alexa Fluor™ 647 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by flow cytometry for its ability to bind anti-human GITR Ligand/TNFSF18 antibody conjugated beads. The concentration of Recombinant Human GITR Ligand/TNFSF18 (GCN4-IZ) Alexa Fluor™ 647 (Catalog # AFR6987) that produces 50% of the binding response is 0.200-2.00 ng/mL.
Specifications
Specifications
| Accession Number | Q9UNG2.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8995 |
| Molecular Weight (g/mol) | M.W.-observed: 20-27 kDa, under reducing conditions., M.W.-theroretical: 19.6 kDa (monomer) |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human GITR Ligand/TNFSF18 protein HA (YPYDVPDYA) GCN4-IZ Human GITR Ligand(Glu52-Ser177)(C-terminus) Accession # Q9UNG2.3 (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction