missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GIP Protein
A cDNA sequence encoding the GIP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11974-10ug
Additional Details : Weight : 0.01000kg
Specifications
GIP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEKKEGHF SALPSLPVGS HAKVSSPQPR GPRYAEGTFI SDYSIAMDKI HQQDFVNWLL AQKGKKNDWK HNITQREARA LELAGQANRK EEEAVEPQSS PAKNPSDEDL LRDLLIQELL ACLLDQTNLC RLRSR | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GIP solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol 0.1M NaCl and 2mM DTT. |