missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GIF Protein

Product Code. 18738937
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18738937

Brand: R&D Systems™ 8387TC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human GIF Protein is derived from HEK293. The Recombinant Human GIF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27352
For Use With (Application) Bioactivity
Gene ID (Entrez) 2694
Molecular Weight (g/mol) Observed M.W.: 44-60kDa, reducing conditions, Theoretical M.W.: 44kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human GIF protein Ser19-Tyr417 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias gastric intrinsic factor (vitamin B synthesis), IF, IFIntrinsic factor, IFMH, IFMHgastric intrinsic factor, INF, TCN3
Common Name GIF
Gene Symbol GIF
Biological Activity Measured by its ability to bind Vitamin B12. The specific binding is =30μg Vitamin B12/mg Recombinant Human GIF.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ser19-Tyr417 with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.