missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein

Product Code. p-7152072
Click to view available options
Quantity:
1 mg
10 mg
5 mg
500 μg
Unit Size:
10mg
1mg
500µg
5mg
This item is not returnable. View return policy

Product Code. 15945959

Brand: enQuireBio™ QP5354500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000506.2
Gene ID (Entrez) 2688
Name GH1 / Growth hormone 1 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol GH1
Biological Activity Human GH1 / Growth hormone 1 induces PDF 9D11 cell proliferation (typical ED50 is 1-15 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF
Buffer 20 mM sodium bicarbonate, pH 8.0
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.