missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein

Product Code. 15945959
Change view
Click to view available options
Quantity:
1 mg
10 mg
5 mg
500 μg
Unit Size:
10mg
1mg
500µg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15945959 500 μg 500µg
15935959 1 mg 1mg
15955959 5 mg 5mg
15925959 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15945959 Supplier enQuireBio™ Supplier No. QP5354500ug
  • ONLINE EXCLUSIVE PRICE
    £213.15
    £205.55 / 500µg
    Save £7.60
    3% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000506.2
Gene ID (Entrez) 2688
Name GH1 / Growth hormone 1 Protein
Quantity 500 μg
Regulatory Status RUO
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol GH1
Biological Activity Human GH1 / Growth hormone 1 induces PDF 9D11 cell proliferation (typical ED50 is 1-15 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF
Buffer 20 mM sodium bicarbonate, pH 8.0
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.