missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein
Click to view available options
Quantity:
1 mg
10 mg
5 mg
500 μg
Unit Size:
10mg
1mg
500µg
5mg
Specifications
Specifications
| Accession Number | NP_000506.2 |
| Gene ID (Entrez) | 2688 |
| Name | GH1 / Growth hormone 1 Protein |
| Quantity | 10 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | GH1 |
| Biological Activity | Human GH1 / Growth hormone 1 induces PDF 9D11 cell proliferation (typical ED50 is 1-15 ng/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction