missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GFAP Protein

Product Code. 18328842
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.50mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328842 1 mg 1mg
18392246 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18328842 Supplier Novus Biologicals™ Supplier No. NBP3119641MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Formulation Lyophilized from 16 mM NaHCO3, containing 0.05% CHAPS and 0.05% Tween 20
Gene ID (Entrez) 2670
Molecular Weight (g/mol) 37.9 kDa
Quantity 1 mg
Storage Requirements Store at -70°C. Avoid freeze-thaw cycles.
Gene Alias ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Gene Symbol GFAP
Reconstitution It is recommended to reconstitute this product with deionized water to its initial concentration.
Purity or Quality Grade >90%, by SDS-PAGE
Protein GFAP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.