missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GEMIN6 Protein

A cDNA sequence encoding the GEMIN6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11952-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928908

  • £155.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GEMIN6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEWMKK GPLEWQDYIY KEVRVTASEK NEYKGWVLTT DPVSANIVLV NFLEDGSMSV TGIMGHAVQT VETMNEGDHR VREKLMHLFT SGDCKAYSPE DLEERKNSLK KWLEKNHIPI TEQGDAPRTL CVAGVLTIDP PYGPENCSSS NEIILSRVQD LIEGHLTASQ
Greater than 90% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GEMIN6 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers