missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDNF Protein

Product Code. 18389437
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18389437 10 μg Lyophilized from a 0.2μm filtered solution in PBS. 10µg
18178540 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10µg
18147400 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50µg
18309267 50 μg Lyophilized from a 0.2μm filtered solution in PBS. 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18389437 Supplier R&D Systems™ Supplier No. 212GD010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ) at 1 μg/mL can bind Recombinant Human GDNF with an apparent Kd <1 nM.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) 11.6 kDa (monomer)
Name GDNF
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human GDNF protein Arg109-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.