missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDNF Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ 212-GD-050/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18309267

  • £1087.44 / 50µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ) at 1 μg/mL can bind Recombinant Human GDNF with an apparent Kd <1 nM.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
11.6 kDa (monomer)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
2668
GDNF
Mouse myeloma cell line,NS0-derived human GDNF protein Arg109-Ile211
Disulfide-linked homodimer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers