missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GDNF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
Brand: R&D Systems™ 212-GD-050
Additional Details : Weight : 0.00970kg
Description
Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ) at 1 μg/mL can bind Recombinant Human GDNF with an apparent Kd <1 nM.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
11.6 kDa (monomer) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2668 | |
GDNF | |
Mouse myeloma cell line,NS0-derived human GDNF protein Arg109-Ile211 | |
Disulfide-linked homodimer | |
Recombinant |