missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-5 Protein

Product Code. 18755497
Click to view available options
:
50ug; Carrier Free
This item is not returnable. View return policy

Product Code. 18755497

Brand: R&D Systems™ 8340G5050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human GDF-5 Protein is derived from E. coli. The Recombinant Human GDF-5 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43026
Formulation Lyophilized from a 0.2μm filtered solution in HCl
Gene ID (Entrez) 8200
Molecular Weight (g/mol) 14kDa
Name GDF-5/BMP-14
Preparation Method Reconstitute at 250μg/mL in 4mM HCl.
Quality Control Testing SDS-PAGE: 14kDa, reducing conditions
Quantity 50μg
Source E. coli-derived human GDF-5/BMP-14 protein Ala382-Arg501
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala382 and Pro383
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.