missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF-15 Protein

A DNA sequence encoding the Homo sapiens (Human) GDF-15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP4998-ye-1mg

 View more versions of this product

Product Code. 15945629

  • £1780.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004855.2
GDF-15 Protein
Research Use Only
GDF15
Recombinant Protein
Yeast
RNGDHCPLGP GRCCRLHTVR ASLEDLGWAD WVLSPREVQV TMCIGACPSQ FRAANMHAQI KTSLHRLKPD TVPAPCCVPA SYNPMVLIQK TDTGVSLQTY DDLLAKDCHC I
Greater than 90% as determined by SDS-PAGE.
9518
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers