missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GDF-15 (Human Cell-expressed) Protein
Click to view available options
Quantity:
10 μg
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
25µg
Description
Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.2-2 ng/mL.
The Recombinant Human GDF-15 (Human Cell-expressed) Protein is derived from HEK293.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 9518 |
| Molecular Weight (g/mol) | 12 kDa (monomer) |
| Name | GDF-15 |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell,HEK293-derived human GDF-15 protein Ala197-Ile308 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction