missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-15 (Human Cell-expressed) Protein

Product Code. 18188150
Click to view available options
Quantity:
10 μg
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18188150

Brand: R&D Systems™ 8146GD010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.2-2 ng/mL.

The Recombinant Human GDF-15 (Human Cell-expressed) Protein is derived from HEK293.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 9518
Molecular Weight (g/mol) 12 kDa (monomer)
Name GDF-15
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human GDF-15 protein Ala197-Ile308
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.