missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-15 (CHO-expressed) Protein

Product Code. 18706276
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18706276

Brand: R&D Systems™ 957GD025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.4-4 ng/mL.

The Recombinant Human GDF-15 Protein is derived from CHO.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 9518
Molecular Weight (g/mol) Predicted MW: 13kDa (monomer) Observed MW: 14kDa, reducing conditions
Name GDF-15
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human GDF-15 protein Ala197-Ile308,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-300μg/mL in sterile 4mM HCI.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.