missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-15, Animal-Free Protein

Product Code. 18775925
Change view
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18775925 50 μg 50µg
18755975 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18775925 Supplier R&D Systems™ Supplier No. Qk0170050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Accession Number Q99988.3
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 9518
Molecular Weight (g/mol) M.W. (Observed): Dimeric GDF-15 protein only, M.W. (Theroretical): 25 kDa (dimer)
Quantity 50 μg
Source E. coli-derived human GDF-15 protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired
Protein GDF-15
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.