missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GASP-1/WFIKKN2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Inhibition Activity
£291.57
Specifications
Gene ID (Entrez) | 124857 |
---|---|
Molecular Weight (g/mol) | 60.7 kDa |
Name | GASP-1/WFIKKNRP |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived human GASP-1/WFIKKN2 protein Leu35-His576,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18354128
|
R&D Systems™
2070-GS-025 |
£291.57
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18177170
|
R&D Systems™
2070-GS-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£291.57
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human GASP-1/WFIKKN2 Protein is derived from NS0. The Recombinant Human GASP-1/WFIKKN2 Protein has been validated for the following applications: Bioactivity, Inhibition Activity.Specifications
124857 | |
GASP-1/WFIKKNRP | |
Mouse myeloma cell line,NS0-derived human GASP-1/WFIKKN2 protein Leu35-His576,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
60.7 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |