missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GARS Protein
A cDNA sequence encoding the GARS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11933-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
GARS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPSPRPVLLR GARAALLLLL PPRLLARPSL LLRRSLSAAS CPPISLPAAA SRSSMDGAGA EEVLAPLRLA VRQQGDLVRK LKEDKAPQVD VDKAVAELKA RKRVLEAKEL ALQPKDDIVD RAKMEDTLKR RFFYDQAFAI YGGVSGLYDF GPVGCALKNN IIQTWRQHFI QEEQILEIDC TMLTPEPVLK TSGHVDKFAD FMVKDVKNGE CFRADHLLKA HLQKLMSDKK CSVEKKSEME SVLAQLDNYG QQELADLFVN YNVKSPITGN DLSPPVSFNL MFKTFIGPGG NMPGYLRPET AQGIFLNFKR LLEFNQGKLP FAAAQIGNSF RNEISPRSGL IRVREFTMAE IEHFVDPSEK DHPKFQNVAD LHLYLYSAKA QVSGQSARKM RLGDAVEQGV INNTVLGYFI GRIYLYLTKV GISPDKLRFR QHMENEMAHY ACDCWDAESK TSYGWIEIVG CADRSCYDLS CHARATKVPL VAEKPLKEPK TVNVVQFEPS KGAIGKAYKK DAKLVMEYLA ICDECYITEM EMLLNEKGEF TIETEGKTFQ LTKDMINVKR FQKTLYVEEV VPNVIEPSFG LGRIMYTVFE HTFHVREGDE QRTFFSFPAV VAPFKCSVLP LSQNQEFMPF VKELSEALTR HGVSHKVDDS SGSIGRRYAR TDEIGVAFGV TIDFDTVNKT PHTATLRDRD SMRQIRAEIS ELPSIVQDLA NGNITWADVE ARYPLFEGQE TGKKETIEE | |
Greater than 80.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GARS is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |