missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAPDH Protein

A DNA sequence encoding the Homo sapiens (Human) GAPDH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8304-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093891

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001243728.1
GAPDH Protein
Research Use Only
GAPDH
Recombinant Protein
E. coli
KVKVGVNGFG RIGRLVTRAA FNSGKVDIVA INDPFIDLNY MVYMFQYDST HGKFHGTVKA ENGKLVINGN PITIFQERDP SKIKWGDAGA EYVVESTGVF TTMEKAGAHL QGGAKRVIIS APSADAPMFV MGVNHEKYDN SLKIISNASC TTNCLAPLAK VIHDNFGIVE GLMTTVHAIT ATQKTVDGPS GKLWRDGRGA LQNIIPASTG AAKAVGKVIP ELNGKLTGMA FRVPTANVSV VDLTCRLEKP AKYDDIKKVV KQASEGPLKG ILGYTEHQVV SSDFNSDTHS STFDAGAGIA LNDHFVKLIS WYDNEFGYSN RVVDLMAHMA SKE
Greater than 90% as determined by SDS-PAGE.
2597
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers