missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Human gamma-Synuclein Active, Monomer Protein

Product Code. 18330755 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
2 x 100 μg Vials
5 x 100 μg Vials
Unit Size:
1 set
100µg
This item is not returnable. View return policy

Product Code. 18330755

Brand: Novus Biologicals NBP314778200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulation PBS pH 7.4
Gene ID (Entrez) 6623
Quantity 2 x 100 μg Vials
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1)
Gene Symbol SNCG
Research Category Alzheimers Research, Cancer, Cellular Markers, Core ESC Like Genes, Mitotic Regulators, Neuroscience, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE
Protein gamma-Synuclein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.