missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GALNT10 Protein
Click to view available options
:
20ug; Carrier Free
Description
Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide MUC5AC-3/13 from AnaSpec, Inc. The specific activity is >180 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q86SR1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 55568 |
| Molecular Weight (g/mol) | M.W. (Observed): 60-68 kDa, reducing conditions; M.W. (theoretical): 62 kDa |
| Name | Polypeptide GalNac Transferase 10/GALNT10 |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human Polypeptide GalNac Transferase 10/GALNT10 protein Leu71-Asn603, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction