Learn More
R&D Systems™ Recombinant Human Galectin-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1339-GA-050
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18106490
  • £345.56 / 50µg
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
The Recombinant Human Galectin-7 Protein is derived from E. coli. The Recombinant Human Galectin-7 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, EDTA, DTT and Trehalose with BSA as a carrier protein.
15 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Recombinant
3963
Galectin-7
E. coli-derived human Galectin-7 protein Ser2-Phe136
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.