missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Galectin-3 protein is fully biologically active when compared to standard. The ED50 as determined by its ability to agglutinate human red blood cells is less than 10 μg/mL.
Specifications
Specifications
| Accession Number | P17931, P17931, P17931 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 1xPBS, 0.02% Tween-20, 3mM beta-ME, pH 7.4. |
| Gene ID (Entrez) | 3958 |
| Molecular Weight (g/mol) | 26 kDa |
| Quantity | 1000 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Galectin-3 as determined by LAL method. |
| Gene Alias | Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?