missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Galectin-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1152-GA-050
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human Galectin-1 Protein is derived from E. coli. The Recombinant Human Galectin-1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. | |
15 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
3956 | |
Galectin-1 | |
E. coli-derived human Galectin-1 protein Ala2-Asp135 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |