missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Galectin 1 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human blood monocytes is in a concentration range of 1.0-10 ng/mL.
Specifications
Specifications
| Accession Number | P09382, P09382, P09382 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS pH 7.0, 3mM beta-Me, 0.02% Tween 20. |
| Gene ID (Entrez) | 3956 |
| Molecular Weight (g/mol) | 14.6 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Galectin-1 as determined by LAL method. |
| Gene Alias | 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?