missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human gAdiponectin/gAcrp30 Protein

Product Code. 18799066
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799066 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799066 Supplier R&D Systems™ Supplier No. 1688AC025
  • ONLINE EXCLUSIVE PRICE
    £371.18
    £329.70 / 25µg
    Save £41.48
    11% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human gAdiponectin/gAcrp30 Protein is derived from NS0. The Recombinant Human gAdiponectin/gAcrp30 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 9370
Molecular Weight (g/mol) 16.7 kDa (monomer)
Name gAdiponectin/gAcrp30
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human gAdiponectin/gAcrp30 protein Ala108-Asn244,with a C-terminal 6-His tag
Storage Requirements 4 months from date of receipt, 2 to 8° C as supplied.
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.