missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAD1 iso1 Protein

Product Code. 15908798
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15908798

Brand: enQuireBio™ QP119181mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GAD1 iso1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 2571
Name GAD1 iso1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol GAD1
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMASSTPS SSATSSNAGA DPNTTNLRPT TYDTWCGVAH GCTRKLGLKI CGFLQRTNSL EEKSRLVSAF KERQSSKNLL SCENSDRDAR FRRTETDFSN LFARDLLPAK NGEEQTVQFL LEVVDILLNY VRKTFDRSTK VLDFHHPHQL LEGMEGFNLE LSDHPESLEQ ILVDCRDTLK YGVRTGHPRF FNQLSTGLDI IGLAGEWLTS TANTNMFTYE IAPVFVLMEQ ITLKKMREIV GWSSKDGDGI FSPGGAISNM YSIMAARYKY FPEVKTKGMA AVPKLVLFTS EQSHYSIKKA GAALGFGTDN VILIKCNERG KIIPADFEAK ILEAKQKGYV PFYVNATAGT TVYGAFDPIQ EIADICEKYN LWLHVDAAWG GGLLMSRKHR HKLNGIERAN SVTWNPHKMM GVLLQCSAIL VKEKGILQGC NQMCAGYLFQ PDKQYDVSYD TGDKAIQCGR HVDIFKFWLM WKAKGTVGFE NQINKCLELA EYLYAKIKNR EEFEMVFNGE PEHTNVCFWY IPQSLRGVPD SPQRREKLHK VAPKIKALMM ESGTTMVGYQ PQGDKANFFR MVISNPAATQ SDIDFLIEEI ERLGQDL
Buffer GAD1 iso1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.