missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human G6PC2 Protein

Product Code. 18425369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425369 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425369 Supplier Novus Biologicals™ Supplier No. H00057818G0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoaffinity Purification
Formulation 25 mM Tris-HCl pH8.0 in 2% glycerol.
Gene ID (Entrez) 57818
Molecular Weight (g/mol) MolecularWeight-theroretical: 39.1 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias EC 3.1.3.9, G6Pase 2, G-6-Pase 2, glucose-6-phosphatase 2, glucose-6-phosphatase, catalytic, 2, IGRPMGC141936, islet-specific G6CP-related protein, Islet-specific glucose-6-phosphatase catalytic subunit-related protein, islet-specific glucose-6-phosphatase-related protein
Gene Symbol G6PC2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein G6PC2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.