missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G1 / S-specific cyclin-D3 Protein
A DNA sequence encoding the Homo sapiens (Human) G1/S-specific cyclin-D3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5778-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129489.1 | |
G1 / S-specific cyclin-D3 Protein | |
Research Use Only | |
CCND3 | |
Recombinant Protein | |
E. coli | |
MELLCCEGTR HAPRAGPDPR LLGDQRVLQS LLRLEERYVP RASYFQCVQR EIKPHMRKML AYWMLEVCEE QRCEEEVFPL AMNYLDRYLS CVPTRKAQLQ LLGAVCMLLA SKLRETTPLT IEKLCIYTDH AVSPRQLRDW EVLVLGKLKW DLAAVIAHDF LAFILHRLSL PRDRQALVKK HAQTFLALCA TDYTFAMYPP SMIATGSIGA AVQGLGACSM SGDELTELLA GITGTEVDCL RACQEQIEAA LRESLREASQ TSSSPAPKAP RGSSSQGPSQ TSTPTDVTAI HL | |
Greater than 80% as determined by SDS-PAGE. |
896 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |