missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G1 / S-specific cyclin-D2 Protein
A DNA sequence encoding the Homo sapiens (Human) G1/S-specific cyclin-D2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5777-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001750.1 | |
G1 / S-specific cyclin-D2 Protein | |
Research Use Only | |
CCND2 | |
Recombinant Protein | |
E. coli | |
MELLCHEVDP VRRAVRDRNL LRDDRVLQNL LTIEERYLPQ CSYFKCVQKD IQPYMRRMVA TWMLEVCEEQ KCEEEVFPLA MNYLDRFLAG VPTPKSHLQL LGAVCMFLAS KLKETSPLTA EKLCIYTDNS IKPQELLEWE LVVLGKLKWN LAAVTPHDFI EHILRKLPQQ REKLSLIRKH AQTFIALCAT DFKFAMYPPS MIATGSVGAA ICGLQQDEEV SSLTCDALTE LLAKITNTDV DCLKACQEQI EAVLLNSLQQ YRQDQRDGSK SEDELDQAST PTDVRDIDL | |
Greater than 80% as determined by SDS-PAGE. |
894 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |