missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human G-CSF Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Acetic Acid.
Unit Size:
25µg
5µg
Description
The Recombinant Human G-CSF Protein is derived from E. coli. The Recombinant Human G-CSF Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Acetic Acid. |
| Gene ID (Entrez) | 1440 |
| Molecular Weight (g/mol) | 18.8 kDa |
| Name | G-CSF |
| Quantity | 5 μg |
| Source | E. coli-derived human G-CSF protein Thr31-Pro204,with an N-terminal Met |
| Storage Requirements | Do not freeze. 6 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction