missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human G-CSF / CSF3 Protein

Product Code. 15905559
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905559 500 μg 500µg
15995549 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15905559 Supplier enQuireBio™ Supplier No. QP2531500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000750.1
Gene ID (Entrez) 1440
Name G-CSF / CSF3 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol CSF3
Biological Activity Shows bioactivity at a concentration of 50 pg/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^7 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.